ENA053246619 Enamine

5-fluoro-3-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD04448734
  • Cas Number : 81718-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12548
Availability details for
Quantity
  • Availability
  • Add to Cart