ENA053246620 Enamine

7-fluoro-3-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD06655338
  • Cas Number : 852388-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12549
Availability details for
Quantity
  • Availability
  • Add to Cart