ENA053246621 Enamine

5-(3-chlorophenyl)-2-(piperidin-1-yl)-[1,3]thiazolo[4,5-b]pyridine-7-carboxylic acid

  • MDL number MFCD06655339
  • Cas Number : 852388-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12550
Availability details for
Quantity
  • Availability
  • Add to Cart