ENA053246624 Enamine

1-[(3-chloro-4-fluorophenyl)sulfonyl]piperazine

  • MDL number MFCD06655342
  • Cas Number : 852388-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12553

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12553
Availability details for
Quantity
  • Availability
  • Add to Cart