ENA053246625 Enamine

5-(chloromethyl)-4-(2-oxopropyl)-8-thia-4,6-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),5-trien-3-one

  • MDL number MFCD05263263
  • Cas Number : 743445-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-133
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12554
Availability details for
Quantity
  • Availability
  • Add to Cart