ENA053246632 Enamine

ethyl[(3-fluoro-4-methoxyphenyl)methyl]amine

  • MDL number MFCD06655347
  • Cas Number : 851903-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12561
Availability details for
Quantity
  • Availability
  • Add to Cart