ENA053246634 Enamine

[3-(morpholin-4-yl)propyl][(pyridin-4-yl)methyl]amine

  • MDL number MFCD01475419
  • Cas Number : 227017-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12563
Availability details for
Quantity
  • Availability
  • Add to Cart