ENA053246640 Enamine

7-amino-2,3-dihydro-1,4-benzodioxine-6-carboxylic acid

  • MDL number MFCD06010010
  • Cas Number : 99358-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12569
Availability details for
Quantity
  • Availability
  • Add to Cart