ENA053246643 Enamine

2-chloro-1-[2-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)pyrrolidin-1-yl]ethan-1-one

  • MDL number MFCD06655353
  • Cas Number : 852217-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12572
Availability details for
Quantity
  • Availability
  • Add to Cart