ENA053246647 Enamine

3-(quinolin-2-yl)propanoic acid

  • MDL number MFCD06655354
  • Cas Number : 39111-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-108
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12576
Availability details for
Quantity
  • Availability
  • Add to Cart