ENA053246648 Enamine

1-isobutyl-4,5-dimethyl-1,3-dihydro-2H-imidazole-2-thione

  • MDL number MFCD06655355
  • Cas Number : 852217-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12577

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12577
Availability details for
Quantity
  • Availability
  • Add to Cart