ENA053246650 Enamine

1-phenyl-2-sulfanyl-1H-imidazole-5-carboxylic acid

  • MDL number MFCD23145179
  • Cas Number : 99361-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245-246
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12579
Availability details for
Quantity
  • Availability
  • Add to Cart