ENA053246669 Enamine

1-(5-tert-butyl-2-methylbenzenesulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD06655368
  • Cas Number : 852400-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12598
Availability details for
Quantity
  • Availability
  • Add to Cart