ENA053246671 Enamine

2-chloro-N-[2-(3-methylphenoxy)ethyl]acetamide

  • MDL number MFCD06655370
  • Cas Number : 50911-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12600
Availability details for
Quantity
  • Availability
  • Add to Cart