ENA053246674 Enamine

4-amino-5-(4-ethoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06655372
  • Cas Number : 156942-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-206
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12603
Availability details for
Quantity
  • Availability
  • Add to Cart