ENA053246683 Enamine

3-(3-ethylphenyl)-2-mercapto-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD06655377
  • Cas Number : 852217-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12612

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12612
Availability details for
Quantity
  • Availability
  • Add to Cart