ENA053246691 Enamine

2-chloro-N-{3-cyano-1-[(furan-2-yl)methyl]-4,5-dimethyl-1H-pyrrol-2-yl}acetamide

  • MDL number MFCD06655382
  • Cas Number : 852217-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12620
Availability details for
Quantity
  • Availability
  • Add to Cart