ENA053246693 Enamine

5-(4-methyl-1,3-thiazol-2-yl)-4-oxo-2-sulfanyl-3-azaspiro[5.5]undec-1-ene-1-carbonitrile

  • MDL number MFCD06660522
  • Cas Number : 852400-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12622
Availability details for
Quantity
  • Availability
  • Add to Cart