ENA053246696 Enamine

N-(2-aminoethyl)-N-pyridin-2-ylamine

  • MDL number MFCD04039672
  • Cas Number : 74764-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45...47
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12625

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-12625
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart