ENA053246700 Enamine

3-(4-isopropylphenyl)-2-mercapto-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD06655388
  • Cas Number : 852400-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12629

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12629
Availability details for
Quantity
  • Availability
  • Add to Cart