ENA053246704 Enamine

5-(chloromethyl)-3-[4-(propan-2-yl)phenyl]-1,2,4-oxadiazole

  • MDL number MFCD06655391
  • Cas Number : 435303-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-36
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12633
Availability details for
Quantity
  • Availability
  • Add to Cart