ENA053246705 Enamine

3-(2,6-dimethylphenyl)-2-mercapto-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD06655392
  • Cas Number : 32262-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12634

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12634
Availability details for
Quantity
  • Availability
  • Add to Cart