ENA053246707 Enamine

2-[(4-ethyl-5-phenyl-4H-1,2,4-triazol-3-yl)sulfanyl]acetic acid

  • MDL number MFCD01163892
  • Cas Number : 305337-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12636
Availability details for
Quantity
  • Availability
  • Add to Cart