ENA053246708 Enamine

ethyl 4-methyl-2-(pentylamino)-1,3-thiazole-5-carboxylate

  • MDL number MFCD06655394
  • Cas Number : 852400-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12637

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12637
Availability details for
Quantity
  • Availability
  • Add to Cart