ENA053246712 Enamine

2-hydrazinyl-3-methyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD01219687
  • Cas Number : 61507-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-124
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12641
Availability details for
Quantity
  • Availability
  • Add to Cart