ENA053246726 Enamine

3-amino-2-methyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD00770876
  • Cas Number : 852399-99-6
  • Molecular Weight : 181.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12655
Availability details for
Quantity
  • Availability
  • Add to Cart