ENA053246730 Enamine

2-chloro-N-(1,2-diphenylethyl)acetamide

  • MDL number MFCD00085848
  • Cas Number : 39787-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12659

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12659
Availability details for
Quantity
  • Availability
  • Add to Cart