ENA053246736 Enamine

5-(2,5-difluorophenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD06655412
  • Cas Number : 956437-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12665
Availability details for
Quantity
  • Availability
  • Add to Cart