ENA053246741 Enamine

2-chloro-1-[1-(ethylsulfonyl)-2,3-dihydro-1H-indol-5-yl]ethanone

  • MDL number MFCD06655416
  • Cas Number : 852399-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12670

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12670
Availability details for
Quantity
  • Availability
  • Add to Cart