ENA053246772 Enamine

1-(3-fluoro-4-methoxybenzyl)piperazine

  • MDL number MFCD05189160
  • Cas Number : 523980-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12701

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12701
Availability details for
Quantity
  • Availability
  • Add to Cart