ENA053246779 Enamine

ethyl 2-[2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]-2-oxoacetate

  • MDL number MFCD06386701
  • Cas Number : 851814-14-7
  • Molecular Weight : 308.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-69
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12708
Availability details for
Quantity
  • Availability
  • Add to Cart