ENA053246783 Enamine

1-[(3,4-dimethoxyphenyl)methyl]piperazine

  • MDL number MFCD00128074
  • Cas Number : 32231-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 27-28
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12712
Availability details for
Quantity
  • Availability
  • Add to Cart