ENA053246785 Enamine

4-chloro-2-[(4-methoxyphenyl)amino]-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD06382012
  • Cas Number : 460730-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12047

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12047
Availability details for
Quantity
  • Availability
  • Add to Cart