ENA053246786 Enamine

1-[4-(propan-2-yl)phenyl]-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD06655442
  • Cas Number : 121690-13-9
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12715
Availability details for
Quantity
  • Availability
  • Add to Cart