ENA053246787 Enamine

cyclopropylthiourea

  • MDL number MFCD06655443
  • Cas Number : 56541-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-143
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12716
Availability details for
Quantity
  • Availability
  • Add to Cart