ENA053246795 Enamine

ethyl 4-methyl-2-[(pentan-2-yl)amino]-1,3-thiazole-5-carboxylate

  • MDL number MFCD06655450
  • Cas Number : 852389-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12724
Availability details for
Quantity
  • Availability
  • Add to Cart