ENA053246797 Enamine

3-(3,4-dimethylphenyl)-2-mercapto-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD03969490
  • Cas Number : 568552-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12726

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12726
Availability details for
Quantity
  • Availability
  • Add to Cart