ENA053246798 Enamine

3-(2-fluorophenyl)-2-mercapto-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD06655453
  • Cas Number : 852389-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12727

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12727
Availability details for
Quantity
  • Availability
  • Add to Cart