ENA053246799 Enamine

2-mercapto-3-(3-methylphenyl)-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD02671329
  • Cas Number : 61388-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12728

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12728
Availability details for
Quantity
  • Availability
  • Add to Cart