ENA053246801 Enamine

3-(3-chlorophenyl)-2-mercapto-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD04048637
  • Cas Number : 42351-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12730

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12730
Availability details for
Quantity
  • Availability
  • Add to Cart