ENA053246802 Enamine

2-mercapto-3-[2-(trifluoromethyl)phenyl]-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD06655456
  • Cas Number : 852389-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12731

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12731
Availability details for
Quantity
  • Availability
  • Add to Cart