ENA053246803 Enamine

3-(2,4-dimethylphenyl)-2-mercapto-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD06655457
  • Cas Number : 852388-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12732

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12732
Availability details for
Quantity
  • Availability
  • Add to Cart