ENA053246807 Enamine

1-(2-methoxyphenyl)-5-phenyl-1H-imidazole-2-thiol

  • MDL number MFCD06655461
  • Cas Number : 852388-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12736
Availability details for
Quantity
  • Availability
  • Add to Cart