ENA053246809 Enamine

1-(2,3-dimethylphenyl)-5-phenyl-1H-imidazole-2-thiol

  • MDL number MFCD06655463
  • Cas Number : 852388-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12738

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12738
Availability details for
Quantity
  • Availability
  • Add to Cart