ENA053246813 Enamine

1-methyl-5-(4-methylphenyl)-1H-imidazole-2-thiol

  • MDL number MFCD04112442
  • Cas Number : 685542-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12742
Availability details for
Quantity
  • Availability
  • Add to Cart