ENA053246816 Enamine

5-(4-bromophenyl)-1-ethyl-1H-imidazole-2-thiol

  • MDL number MFCD06655469
  • Cas Number : 852388-98-8
  • Molecular Weight : 283.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12745
Availability details for
Quantity
  • Availability
  • Add to Cart