ENA053246817 Enamine

1-ethyl-5-(4-methylphenyl)-1H-imidazole-2-thiol

  • MDL number MFCD06655470
  • Cas Number : 852388-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12746

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12746
Availability details for
Quantity
  • Availability
  • Add to Cart