ENA053246819 Enamine

5-(2,3-dihydro-1,4-benzodioxin-6-yl)-1-ethyl-1H-imidazole-2-thiol

  • MDL number MFCD06655472
  • Cas Number : 852388-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12748

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12748
Availability details for
Quantity
  • Availability
  • Add to Cart