ENA053246822 Enamine

N-(1-benzyl-3-cyano-4,5-dimethyl-1H-pyrrol-2-yl)-2-chloroacetamide

  • MDL number MFCD06655475
  • Cas Number : 852388-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12751
Availability details for
Quantity
  • Availability
  • Add to Cart