ENA053246823 Enamine

1-pyridin-3-ylethanamine

  • MDL number MFCD01691641
  • Cas Number : 56129-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12752

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-12752
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart